Learn More
OraSure Technologies OraQuick™ Advance™ Rapid HIV-1/2 Antibody Test
HIV Testing anytime, anywhere, made easy. The OraQuick™ ADVANCE™ Rapid HIV-1/2 Antibody Test is an easy-to-use rapid HIV test that offers several specimen types - oral fluid, fingerstick whole blood, venipuncture whole blood and plasma. Yielding accurate results in as little as 20 minutes.
$493.00 - $1969.00
Specifications
Certifications/Compliance ISO 13485 Certified. FDA approved under PMA BP010047. Inspected to 21 CFR 820 and MDSAP certification.
Content And Storage 2°C to 27°C
Description Detects HIV-1 and HIV-2. It is developed with synthetic peptides that represent the envelope region.
CE Marker Yes
Detectable Analytes HIV-1/2
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-222-100
View Documents
OraSure Technologies
10010079
25 Tests
Pack of 25 for $493.00
Edge
Add to Cart
23-222-101
View Documents
OraSure Technologies
10010078
100 Tests
Pack of 100 for $1,969.00
Edge
Add to Cart
Description
Description
  • The OraQuick™ ADVANCE™ Rapid HIV-1/2 Antibody Test detects antibodies to HIV-1 and HIV-2 in 20 minutes.
  • Approved for oral fluid, plasma, fingerstick or venipuncture whole blood specimens.
  • CLIA-waived for oral fluid, fingerstick and venipuncture whole blood and offers the ability to test in non-traditional testing environments, such as outreach programs and mobile testing clinics.
  • CLIA Moderately Complex for plasma samples.
  • The OraQuick™ ADVANCE™ is comprised of a single-use test device and a single-use vial containing a pre-measured amount of a buffered developer solution.
  • Each component is sealed in separate compartments of a single pouch to form the test.
  • 25 Test count packaging includes:  (25) Divided Pouches with Test Device and 1 mL Vial Developer Solution, (5) Reusable Test Stands, (25) Specimen Collection Loops, (25) Information Pamphlets, Package Insert, Customer Letter.
  • 100 Test count packaging includes: (100) Divided pouches with (1) test device, (1) absorbent pack, (1) 1 mL developer solution vial; (10) reusable test stands, (100) specimen collection loops, (100) subject information pamphlets, (1) package insert, (1) customer letter.
  • Store unused tests at 2 to 27°C (36-80°F).
  • If stored refrigerated, ensure that the divided pouch is brought to operating temperature (15-37°C, 59-99°F) before opening.
Specifications
Specifications
ISO 13485 Certified. FDA approved under PMA BP010047. Inspected to 21 CFR 820 and MDSAP certification.
Detects HIV-1 and HIV-2. It is developed with synthetic peptides that represent the envelope region.
HIV-1/2
2°C to 27°C
Yes
Yes