Learn More
Cole-Parmer Hirschmann opus Dispenser, 20 mL, 100 to 240 VAC
Supplier:  Cole-Parmer 2550302
Encompass_Preferred
The motorized liquid dispenser with a reagent recirculation system provides accurate and reliable serial dispensing results up to 9999 times. The dispensing procedure is configured on an external control module featuring a user-friendly IFT touch screen. All tasks are easily and quickly configured on the main screen; up to nine different user-defined programs can be saved in memory and reactivated at any time. The last setting is automatically saved when unit is turned off. Unique stepper function allows you to dispense several different volumes from a single cylinder filling. Recirculation system makes it possible for rapid venting when priming the pump without loss of reagent. Universal luer connection lets you to add filters and drying tubes to the discharge path. Dispenser rotates freely on the bottle, providing flexibility. Quick-Cal function allows you to calibrate the dispenser for different temperatures and reagents.
Catalog No. NC1857734