Learn More
MilliporeSigma™ Opticap™ Sterile XL300 Capsule Filters
For sterilizing grade performance and mycoplasma removal
Supplier:  MilliporeSigma™ KHVES003HH3
 View more versions of this product
Catalog No. KHVES003HH3
Add to Cart
Description
Description
  • High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
  • On-board PES membrane prefilter for extended throughput in high fouling media
  • Robust construction
  • Withstand multiple sterilization cycles
  • Easy wetting and integrity testing
  • Broad chemical compatibility across a wide pH range (pH 1–14)
  • Hydrophilic Polyethersulfone filter
  • Polyester support
  • Structural Components: Gamma stable polypropylene/Polysulfone
  • Silicone vent O-rings
  • Gamma compatible sterilization
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Specifications
Specifications
0.5/0.1 μm
0.03 m2
0.56 in. (14 mm) Hose Barb
4.7 in.
Sterile
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
0.56 in. (14mm) Hose Barb
11.9 cm
Product Suggestions
Product Suggestions