Learn More
Optimize Technologies OPTI-LYNX™ All-In-One Column Kit with Guard Column Holder ( Includes: 2 fittings, 2 bayonet connectors, 2 plugs, 2 holder caps, sealing element & holder tube, guard holder tube)
SDP
Supplier:  Optimize Technologies 1102323AA
Encompass_Preferred
The OPTI-LYNX™ Guard Column is a high-performance, cartridge-based guard system, suitable for use with any 10-32 analytical column. OPTI-LYNX combines effective column protection with the convenience of quick-connect operation .Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2μL. You get quick-connect convenience and low dispersion, all in one package.
Catalog No. 50-113-454