Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter: Probes



For use with Open Source Bio (OSB) transmitter.
$22.00 - $360.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
S21160
![]() ![]() |
Boekel Scientific™
9510001/EMD |
pH Probe |
Each for $122.00
|
|
S21161
![]() ![]() |
Boekel Scientific™
9510002/EMD |
Conductivity Probe (Type k=1.0) |
Each for $164.00
|
|
S21162
![]() ![]() |
Boekel Scientific™
9510003/EMD |
DO Probe |
Each for $360.00
|
|
S21163
![]() ![]() |
Boekel Scientific™
9510004/EMD |
Temperature Probe |
Each for $22.00
|
|
Description
For use with Open Source Bio (OSB) transmitter.Specifications
Open Source Bio (OSB) Transmitter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title