Learn More
Design Point Solutions ONYX FILAMENT SPOOL (800 CM3)
Supplier: Design Point Solutions FMF0001
Onyx Spools, High-Performance Filament.Our Onyx material is at the core of all Markforged printers starting with the Onyx One. It is a beautiful black filament made from combining tough nylon with micro-carbon reinforcement. Onyx gives you stiff and dimensionally stable engineering grade parts, with twice the strength of other 3D printed plastics. Onyx parts have a high quality surface finish and high heat tolerance.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.