Learn More
Dunlop™ Onguard™ Superpoly Boots with Chevron Sole
SDP
Ideal for use in food processing, especially red-meat processing and petrochemical applications.
Supplier:  Dunlop™ 853241000
 View more versions of this product
Catalog No. 19-024-358
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Waterproof with a high degree of chemical resistance
  • Steel toe and shank provide optimal protection
  • Chevron outsole provides excellent slip resistance
  • Pull-up and kick-off lugs
  • Removable or replaceable preformed insole for added comfort
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications
Specifications
Specifications
Red
10
Steel
Chemicals
16 in.
Chevron outsole
Men's
Knee Boot
Product Suggestions
Product Suggestions