Learn More
Dunlop™ Onguard™ Superpoly Boots with Chevron Sole
Ideal for use in food processing, especially red-meat processing and petrochemical applications.
Supplier: Dunlop™ 853240800
This item is not returnable.
View return policy
Description
- Waterproof with a high degree of chemical resistance
- Steel toe and shank provide optimal protection
- Chevron outsole provides excellent slip resistance
- Pull-up and kick-off lugs
- Removable or replaceable preformed insole for added comfort
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications
Specifications
ASTM F2413-05, ANSI Z41 PT99 M I/75 C/75 PR | |
Pull-on | |
16 in. | |
Urethane, PVC | |
8 | |
Steel |
Red | |
Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants | |
Kick-off | |
Steel | |
Tan Chevron outsole and replaceable cushion insoles | |
Chemicals |