Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dunlop™ Onguard™ Men's Standard Cushion Insoles
$8.05
Specifications
Color | White |
---|---|
For Use With (Application) | For comfort and protection for protective footwear |
Material | Foam |
Gender | Men's |
Product Type | Standard Cushion Insole |
Specifications
White | |
Foam | |
Standard Cushion Insole |
For comfort and protection for protective footwear | |
Men's |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title