Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Green, 488
Suitable for in situ apoptosis detection of tissue samples (Paraffin embedding, freezing and ultrathin section) and cells (Cell smears, cell climbing smears).
$271.59 - $635.23
Specifications
Content And Storage | Store at -20C. |
---|---|
Description | OneStep TUNEL |
For Use With (Application) | Flow Cytometry, Fluorescence Imaging |
Product Type | Apoptosis Kit |
Specifications
Store at -20C. | |
OneStep TUNEL | |
Flow Cytometry, Fluorescence Imaging | |
Apoptosis Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title