Learn More
Clontech Labs 3P ONE SHOT LA PCR MIX 1 SET
SDP
Supplier:  Clontech Labs 3P RR004
Encompass
PCR Reagent; Takara; One-Shot LA Taq DNA Polymerase PCR Mix; 24 x 25uL; Contains polymerase, reaction buffer, and dNTPs at a 2X concentration; 25uL aliquots are predispensed into 0.2mL microcentrifuge tubes
Catalog No. 50-443-977