Learn More
Hach Company One reagent stock solution, 500 mL
Supplier: Hach Company 9459400
QBD1200 concentrated reagent solution contains phosphoric acid and ammonium persulfate. For TOC determinations.