Learn More
Diba Omnifit™ Replacement Filters for Glass Bottles
Solvent frits and filters for the bottom of the bottle remove even small particulates
$86.00
Catalog Number | Mfr. No. | For Use With (Equipment) | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | No. per Case | Price | Quantity & Availability | ||||
OF9F45
|
Diba
2194334 |
GL45 Bottle | Each |
Each for $86.00
|
|
||||
OF9FE45
|
Diba
2194338 |
GL45 Bottle | 20/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- All-PTFE construction enables the filters to be used with harsh chemicals
- The filter units have slots in the body to enable virtually all liquid to be removed from the bottle, minimizing waste
Specifications
PTFE | |
10 μm | |
Replacement Filter |