Learn More
Diba Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ SolventPlus™ Columns
$306.00 - $1704.00
Specifications
Type | Liquid Chromatography Column |
---|---|
For Use With (Equipment) | Omnifit™ EZ SolventPlus™ Columns |
Material | PTFE |
Catalog Number | Mfr. No. | Description | Endcapped | Diameter (Metric) Inner | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Endcapped | Diameter (Metric) Inner | Price | Quantity & Availability | |||
14-533-233
|
Diba
1194259 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 6.6mm; 1/ea | Adjustable | 6.6 mm |
Each for $608.00
|
|
|||
14-533-234
|
Diba
1194260 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 10mm; 1/ea | Adjustable | 10 mm |
Each for $727.00
|
|
|||
14-533-235
|
Diba
1194261 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 15mm; 1/ea | Adjustable | 15 mm |
Each for $845.00
|
|
|||
14-533-236
|
Diba
1194262 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 25mm; 1/ea | Adjustable | 25 mm |
Each for $968.00
|
|
|||
14-533-237
|
Diba
1194263 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 35mm; 1/ea | Adjustable | 35 mm |
Each for $1,117.00
|
|
|||
14-533-238
|
Diba
1194264 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Adjustable Column Endpiece, 50mm; 1/ea | Adjustable | 50 mm |
Each for $1,704.00
|
|
|||
14-533-239
|
Diba
1194266 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 6.6mm; 1/ea | Fixed | 6.6 mm |
Each for $306.00
|
|
|||
14-533-240
|
Diba
1194267 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 10mm; 1/ea | Fixed | 10 mm |
Each for $339.00
|
|
|||
14-533-241
|
Diba
1194268 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 15mm; 1/ea | Fixed | 15 mm |
Each for $430.00
|
|
|||
14-533-242
|
Diba
1194269 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 25mm; 1/ea | Fixed | 25 mm |
Each for $485.00
|
|
|||
14-533-243
|
Diba
1194270 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 35mm; 1/ea | Fixed | 35 mm |
Each for $788.00
|
|
|||
14-533-244
|
Diba
1194271 |
Omnifit™ EZ SolventPlus™ EZ SolventPlus Fixed Column Endpiece, 50mm; 1/ea | Fixed | 50 mm |
Each for $957.00
|
|
|||
Specifications
Liquid Chromatography Column | |
Omnifit™ EZ SolventPlus™ Columns | |
PTFE |