Learn More
Diba Omnifit™ O-Rings for Chromatography Columns
Select from FKM or FFKM (Chemraz™) Omnifit™ O-Rings for Chromatography Columns
$18.50 - $790.00
Specifications
Type | Liquid Chromatography Column |
---|---|
For Use With (Equipment) | Omnifit™ HiT Chromatography Columns |
Quantity | 6 |
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Description | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Description | Material | Price | Quantity & Availability | |||
14-533-152
|
Diba
1194544 |
3 mm | Column O-Ring, 3mm, FKM; 6/pk | FKM |
Pack of 6 for $18.50
|
|
|||
14-533-153
|
Diba
1194545 |
6.6 mm | Column O-Ring, 6.6mm, FFKM; 2/pk | FFKM |
Pack of 2 for $220.00
|
|
|||
14-533-154
|
Diba
1194546 |
6.6 mm | Column O-Ring, 6.6mm, FKM; 10/pk | FKM |
Pack of 10 for $19.50
|
|
|||
14-533-155
|
Diba
1194547 |
10 mm | Column O-Ring, 10mm, FFKM; 2/pk | FFKM |
Pack of 2 for $248.00
|
|
|||
14-533-156
|
Diba
1194548 |
10 mm | Column O-Ring, 10mm, FKM; 10/pk | FKM |
Pack of 10 for $24.75
|
|
|||
14-533-157
|
Diba
1194549 |
15 mm | Column O-Ring, 15mm, FFKM; 2/pk | FFKM |
Pack of 2 for $253.00
|
|
|||
14-533-159
|
Diba
1194551 |
25 mm | Column O-Ring, 25mm, FFKM; 2/pk | FFKM |
Pack of 2 for $337.00
|
|
|||
14-533-160
|
Diba
1194552 |
25 mm | Column O-Ring, 25mm, FKM; 6/pk | FKM |
Pack of 6 for $35.50
|
|
|||
14-533-161
|
Diba
1194553 |
35 mm | Column O-Ring, 35mm, FFKM; 2/pk | FFKM |
Pack of 2 for $394.00
|
|
|||
14-533-163
|
Diba
1194555 |
50 mm | Column O-Ring, 50mm, FFKM; 2/pk | FFKM |
Pack of 2 for $790.00
|
|
|||
Specifications
Liquid Chromatography Column | |
Omnifit™ HiT Chromatography Columns | |
6 |