Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
$1061.00 - $3517.00
Specifications
Type | Liquid Chromatography Column |
---|---|
Temperature (Metric) Range | 4 to 40°C |
Pore Size | 10 and 30 μm |
Material | Borosilicate Glass, PTFE, Chemraz™ |
Max. Temperature | 40°C |
Catalog Number | Mfr. No. | Column Capacity | Description | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Column Capacity | Description | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||
14-533-006
|
Diba
1194398 |
4.8 to 10.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 330mm; 1/ea | 6.6 mm | 330 mm |
Each for $1,322.00
|
|
||
14-533-036
|
Diba
1194428 |
10.6 to 38.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 15 x 250mm; 1/ea | 15 mm | 250 mm |
Each for $1,492.00
|
|
||
14-533-066
|
Diba
1194458 |
2.3 to 115.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 35 x 150mm; 1/ea | 35 mm | 150 mm |
Each for $2,526.00
|
|
||
14-533-054
|
Diba
1194446 |
29.5 to 108.0 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 25 x 250mm; 1/ea | 25 mm | 250 mm |
Each for $1,871.00
|
|
||
14-533-021
|
Diba
1194413 |
11 to 23.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 10 x 330mm; 1/ea | 10 mm | 330 mm |
Each for $1,382.00
|
|
||
14-533-051
|
Diba
1194443 |
1.2 to 58.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 25 x 150mm; 1/ea | 25 mm | 150 mm |
Each for $1,727.00
|
|
||
14-533-081
|
Diba
1194473 |
280.4 to 600.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 50 x 330mm; 1/ea | 50 mm | 330 mm |
Each for $3,517.00
|
|
||
14-533-084
|
Diba
1194476 |
420.6 to 741.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 50 x 400mm; 1/ea | 50 mm | 400 mm |
Each for $3,365.00
|
|
||
14-533-015
|
Diba
1194407 |
0.2 to 9.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 10 x 150mm; 1/ea | 10 mm | 150 mm |
Each for $1,554.00
|
|
||
14-533-045
|
Diba
1194437 |
54.8 to 83.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 15 x 500mm; 1/ea | 15 mm | 500 mm |
Each for $1,549.00
|
|
||
14-533-075
|
Diba
1194467 |
202.0 to 356.0 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 35 x 400mm; 1/ea | 35 mm | 400 mm |
Each for $2,469.00
|
|
||
14-533-362
|
Diba
1194389 |
0.1 to 2.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 100mm; 1/ea | 6.6 mm | 100 mm |
Each for $1,080.00
|
|
||
14-533-027
|
Diba
1194419 |
24.3 to 36.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 10 x 500mm; 1/ea | 10 mm | 500 mm |
Each for $1,284.00
|
|
||
14-533-057
|
Diba
1194449 |
68.7 to 147.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 25 x 330mm; 1/ea | 25 mm | 330 mm |
Each for $1,858.00
|
|
||
14-533-087
|
Diba
1194479 |
620.9 to 941.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 50 x 500mm; 1/ea | 50 mm | 500 mm |
Each for $3,422.00
|
|
||
14-533-018
|
Diba
1194410 |
4.7 to 17.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 10 x 250mm; 1/ea | 10 mm | 250 mm |
Each for $1,477.00
|
|
||
14-533-048
|
Diba
1194440 |
1.2 to 34.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 25 x 100mm; 1/ea | 25 mm | 100 mm |
Each for $1,754.00
|
|
||
14-533-078
|
Diba
1194470 |
120.2 to 440.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 50 x 250mm; 1/ea | 50 mm | 250 mm |
Each for $3,403.00
|
|
||
14-533-009
|
Diba
1194401 |
7.2 to 12.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 400mm; 1/ea | 6.6 mm | 400 mm |
Each for $1,095.00
|
|
||
14-533-039
|
Diba
1194431 |
24.7 to 53.0 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 15 x 330mm; 1/ea | 15 mm | 330 mm |
Each for $1,567.00
|
|
||
14-533-069
|
Diba
1194461 |
27.7 to 211.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 35 x 250mm; 1/ea | 35 mm | 250 mm |
Each for $2,527.00
|
|
||
14-533-000
|
Diba
1194392 |
0.1 to 4.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 150mm; 1/ea | 6.6 mm | 150 mm |
Each for $1,075.00
|
|
||
14-533-030
|
Diba
1194422 |
0.4 to 12.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 15 x 100mm; 1/ea | 15 mm | 100 mm |
Each for $1,490.00
|
|
||
14-533-060
|
Diba
1194452 |
103.1 to 181.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 25 x 400mm; 1/ea | 25 mm | 400 mm |
Each for $1,824.00
|
|
||
14-533-012
|
Diba
1194404 |
0.2 to 5.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 10 x 100mm; 1/ea | 10 mm | 100 mm |
Each for $1,486.00
|
|
||
14-533-042
|
Diba
1194434 |
37.1 to 65.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 15 x 400mm; 1/ea | 15 mm | 400 mm |
Each for $1,507.00
|
|
||
14-533-072
|
Diba
1194464 |
134.7 to 288.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 35 x 330mm; 1/ea | 35 mm | 330 mm |
Each for $2,326.00
|
|
||
14-533-003
|
Diba
1194395 |
2.1 to 7.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 250mm; 1/ea | 6.6 mm | 250 mm |
Each for $1,075.00
|
|
||
14-533-033
|
Diba
1194425 |
0.4 to 21.2 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 15 x 150mm; 1/ea | 15 mm | 150 mm |
Each for $1,477.00
|
|
||
14-533-063
|
Diba
1194455 |
152.2 to 230.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 25 x 500mm; 1/ea | 25 mm | 500 mm |
Each for $1,792.00
|
|
||
14-533-359
|
Diba
1194386 |
0.1 to 0.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 6.6 x 50mm; 1/ea | 6.6 mm | 50 mm |
Each for $1,061.00
|
|
||
14-533-024
|
Diba
1194416 |
16.5 to 29.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 10 x 400mm; 1/ea | 10 mm | 400 mm |
Each for $1,315.00
|
|
||
Description
- PEEK™ column threads offer outstanding durability
- Ergonomically designed adjustable endpiece offers quick and easy height adjustment
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Liquid Chromatography Column | |
10 and 30 μm | |
40°C |
4 to 40°C | |
Borosilicate Glass, PTFE, Chemraz™ | |
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |