Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces feature PTFE, borosilicate glass, and Chemraz™ wetted materials for exceptional chemical resistance
$800.00 - $2946.00
Specifications
Type | Liquid Chromatography Column |
---|---|
Temperature (Metric) Range | 4 to 40°C |
Pore Size | 10 and 30 μm |
Material | Borosilicate Glass, PTFE, Chemraz™ |
Max. Temperature | 40°C |
Catalog Number | Mfr. No. | Column Capacity | Description | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Column Capacity | Description | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||
14-533-016
|
Diba
1194408 |
3.1 to 9.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 150mm; 1/ea | 10 mm | 150 mm |
Each for $1,128.00
|
|
||
14-533-046
|
Diba
1194438 |
68.9 to 83.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 500mm; 1/ea | 15 mm | 500 mm |
Each for $1,211.00
|
|
||
14-533-076
|
Diba
1194468 |
279.0 to 356.0 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 35 x 400mm; 1/ea | 35 mm | 400 mm |
Each for $2,116.00
|
|
||
14-533-007
|
Diba
1194399 |
7.5 to 10.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 330mm; 1/ea | 6.6 mm | 330 mm |
Each for $966.00
|
|
||
14-533-037
|
Diba
1194429 |
24.7 to 38.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 250mm; 1/ea | 15 mm | 250 mm |
Each for $1,180.00
|
|
||
14-533-067
|
Diba
1194459 |
38.5 to 115.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 35 x 150mm; 1/ea | 35 mm | 150 mm |
Each for $2,231.00
|
|
||
14-533-363
|
Diba
1194390 |
0.1 to 2.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 100mm; 1/ea | 6.6 mm | 100 mm |
Each for $925.00
|
|
||
14-533-028
|
Diba
1194420 |
30.6 to 36.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 500mm; 1/ea | 10 mm | 500 mm |
Each for $1,023.00
|
|
||
14-533-058
|
Diba
1194450 |
108.0 to 147.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 330mm; 1/ea | 25 mm | 330 mm |
Each for $1,407.00
|
|
||
14-533-088
|
Diba
1194480 |
781.2 to 941.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 50 x 500mm; 1/ea | 50 mm | 500 mm |
Each for $2,946.00
|
|
||
14-533-019
|
Diba
1194411 |
11.0 to 17.3 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 250mm; 1/ea | 10 mm | 250 mm |
Each for $945.00
|
|
||
14-533-049
|
Diba
1194441 |
1.2 to 34.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 100mm; 1/ea | 25 mm | 100 mm |
Each for $1,393.00
|
|
||
14-533-079
|
Diba
1194471 |
280.4 to 440.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 50 x 250mm; 1/ea | 50 mm | 250 mm |
Each for $2,822.00
|
|
||
14-533-010
|
Diba
1194402 |
9.9 to 12.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 400mm; 1/ea | 6.6 mm | 400 mm |
Each for $1,041.00
|
|
||
14-533-040
|
Diba
1194432 |
38.9 to 53.0 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 330mm; 1/ea | 15 mm | 330 mm |
Each for $1,219.00
|
|
||
14-533-070
|
Diba
1194462 |
134.7 to 211.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 35 x 250mm; 1/ea | 35 mm | 250 mm |
Each for $2,079.00
|
|
||
14-533-022
|
Diba
1194414 |
17.3 to 23.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 330mm; 1/ea | 10 mm | 330 mm |
Each for $1,085.00
|
|
||
14-533-052
|
Diba
1194444 |
19.6 to 58.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 150mm; 1/ea | 25 mm | 150 mm |
Each for $1,476.00
|
|
||
14-533-082
|
Diba
1194474 |
440.7 to 600.9 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 50 x 330mm; 1/ea | 50 mm | 330 mm |
Each for $2,774.00
|
|
||
14-533-013
|
Diba
1194405 |
0.2 to 5.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 100mm; 1/ea | 10 mm | 100 mm |
Each for $1,118.00
|
|
||
14-533-043
|
Diba
1194435 |
51.2 to 65.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 400mm; 1/ea | 15 mm | 400 mm |
Each for $1,196.00
|
|
||
14-533-073
|
Diba
1194465 |
211.7 to 288.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 35 x 330mm; 1/ea | 35 mm | 330 mm |
Each for $2,155.00
|
|
||
14-533-004
|
Diba
1194396 |
4.8 to 7.5 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 250mm; 1/ea | 6.6 mm | 250 mm |
Each for $800.00
|
|
||
14-533-034
|
Diba
1194426 |
7.1 to 21.2 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 150mm; 1/ea | 15 mm | 150 mm |
Each for $1,247.00
|
|
||
14-533-064
|
Diba
1194456 |
191.4 to 230.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 500mm; 1/ea | 25 mm | 500 mm |
Each for $1,413.00
|
|
||
14-533-001
|
Diba
1194393 |
1.4 to 4.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 150mm; 1/ea | 6.6 mm | 150 mm |
Each for $860.00
|
|
||
14-533-031
|
Diba
1194423 |
0.4 to 12.4 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 15 x 100mm; 1/ea | 15 mm | 100 mm |
Each for $1,234.00
|
|
||
14-533-061
|
Diba
1194453 |
142.4 to 181.6 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 400mm; 1/ea | 25 mm | 400 mm |
Each for $1,432.00
|
|
||
14-533-360
|
Diba
1194387 |
0.1 to 0.7 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 50mm; 1/ea | 6.6 mm | 50 mm |
Each for $906.00
|
|
||
14-533-025
|
Diba
1194417 |
22.8 to 29.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 10 x 400mm; 1/ea | 10 mm | 400 mm |
Each for $1,113.00
|
|
||
14-533-055
|
Diba
1194447 |
68.7 to 108.0 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 25 x 250mm; 1/ea | 25 mm | 250 mm |
Each for $1,287.00
|
|
||
14-533-085
|
Diba
1194477 |
580.9 to 741.1 mL | Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 50 x 400mm; 1/ea | 50 mm | 400 mm |
Each for $2,869.00
|
|
||
Description
- PEEK™ column threads offer outstanding durability
- Ergonomically designed adjustable endpiece offers quick and easy height adjustment
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Liquid Chromatography Column | |
10 and 30 μm | |
40°C |
4 to 40°C | |
Borosilicate Glass, PTFE, Chemraz™ | |
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |