Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Columns with 2 Fixed Endpieces
Omnifit™ Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system.
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11-100-9256
|
MilliporeSigma™ Supelco™
K5128U |
6.6 mm | 250 mm |
Each for $433.00
|
N/A | ||||
11-100-9126
|
MilliporeSigma™ Supelco™
K5130U |
6.6 mm | 400 mm |
Each for $472.00
|
N/A | ||||
11-100-9087
|
MilliporeSigma™ Supelco™
K5134U |
10 mm | 330 mm |
Each for $484.00
|
N/A | ||||
11-100-9088
|
MilliporeSigma™ Supelco™
K5135U |
10 mm | 400 mm |
Each for $484.00
|
N/A | ||||
11-100-8959
|
MilliporeSigma™ Supelco™
K5136U |
10 mm | 500 mm |
Each for $533.00
|
N/A | ||||
11-100-8843
|
MilliporeSigma™ Supelco™
K5140U |
15 mm | 330 mm |
Each for $608.00
|
N/A | ||||
11-100-8794
|
MilliporeSigma™ Supelco™
K5141U |
15 mm | 400 mm |
Each for $626.00
|
N/A | ||||
11-100-8747
|
MilliporeSigma™ Supelco™
K5142U |
15 mm | 500 mm |
Each for $649.00
|
N/A | ||||
11-100-8620
|
MilliporeSigma™ Supelco™
K5146U |
25 mm | 330 mm |
Each for $723.00
|
N/A | ||||
11-100-8633
|
MilliporeSigma™ Supelco™
K5147U |
25 mm | 400 mm |
Each for $716.00
|
N/A | ||||
11-100-8550
|
MilliporeSigma™ Supelco™
K5148U |
25 mm | 500 mm |
Each for $768.00
|
N/A | ||||
Description
This configuration of Benchmark columns is non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.Specifications
Omnifit | |
Chromatography |
Omnifit is a registered trademark of Diba Industries