Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Omeprazole, Tocris Bioscience™
Click to view available options
Quantity:
50mg
Description
H+,K+-ATPase inhibitor (IC50 = 5.8 μM) that displays antisecretory and antiulcer activity. Inhibits gastric acid secretion (IC50 = 0.16 μM for histamine-induced acid formation) and reduces gastric lesion formation induced by a variety of ulcerative stimuli. Antibacteral against Helicobacter pylori in vitro. Also inhibits CYP2C19, CYP2C9 and CYP3A (Ki values are 3.1, 40.1 and 84.4 μM respectively) and blocks swelling-dependent chloride channels (ICIswell). Enhances anti-SARS-CoV-2 acitvity of Remdesivir and Aprotinin (Cat. No. 4139).
Specifications
Quantity | 50mg |
Formula Weight | 345.42 |
Percent Purity | >99% |
Chemical Name or Material | Omeprazole |
Safety and Handling
Recommended Storage : Desiccate at +4°C