Learn More
Cytiva (Formerly Pall Lab) Nanosep Centrifugal Devices with Omega™ Membrane - 30K, red (500/pkg)
Supplier: Cytiva OD030C35
Fits standard centrifuge rotors that accept 1.5 mL tubes This product is not approved for use in medical, clinical, surgical or other patient applications.