Learn More
Cayman Chemical Oligomycin A, 579-13-5, 1 mg
Supplier: Cayman Chemical 113421
An inhibitor of mitochondrial F1FO ATP synthase that induces apoptosis in a variety of cell types (average GI50 = 270 nM); exhibits antifungal, antitumor, and nematocidal activities, but has poor solubility in water and other biocompatible solvents