Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OK-1™ Half-Finger Anti-Vibration Gloves
Click to view available options
Size:
Medium
Large
Description
- A precurved shape and half finger design provide dexterity
- Gloves offer superior protection from sources of vibration and impact
- Maintain a consistent grip due to the curved design that forms to the natural shape of the hand
- Machine washable
Specifications
Color | Brown |
Hand | Specific |
Material | Grain Cowhide Leather |
Product Type | Work Gloves |
Size | Medium |
For Use With (Application) | Construction, Machine Operation, and Manufacturing |
Series | OK980 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction