Learn More
Parker domnick hunter™ Oil-XPlus™ Filter Elements for Gas Generators
Replacement elements for >Oil-XPlus Compressed Air Filters used with Zero Air Generators
Supplier: Parker domnick hunter™ K009AA
Description
- Use specially treated borosilicate glass microfibers to repel oil and water, reducing pressure drop and minimizing running costs
- Comply with ISO 8573-1 Air Quality Class 1.3.1
- Maximum usage: 8000 hours
Specifications
Outlet Filter Element | |
ISO 8573-1 | |
Uhp-10za and uhp-35za |