Learn More
New Pig Oil-Only Absorbent Mat Roll
Lasts 2x longer than ordinary mats for fewer change-outs, while only absorbing oil and fuels, NOT water. Loves oil. Hates water.
Supplier: New Pig MAT401
This item is not returnable.
View return policy
Description
- Our most cost-effective mat when you only need to absorb oil-based liquids
- Multiple layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
- Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
- Highly absorbent, fine-fiber construction won't leave behind oil or fiber residue Absorbs oil and fuels, NOT water
- White color tells you it's only for oils and fuels; draws attention to machine leaks and clearly shows saturation level during spill response
- Floats to clean up spills on water
- Can be wrung out and incinerated after use to reduce waste or for fuels blending
- Rolls are ideal for covering large areas and soaking up leaks and overspray
- Easy-tear perforations let you take only what you need, so you use less mat and save money
- Heavy-weight construction is highly durable and absorbent for demanding tasks and large-volume cleanups
Specifications
Oil Absorbent Roll |