Learn More
Showa Denko America Inc OHpak SB-803 HQ
Supplier: Showa Denko America Inc F6429102
Polyhydroxymethacrylate packed columns for aqueous SEC (GFC) analysis, supports a wide range of molecular weight sample analysis, the eluent can be replaced with DMF (except SB-802 HQ and SB-807 HQ), enabling the analysis of polar polymers, SB-804 and SB-805 used for accurate MW determination of gelatin by PAGI method, USP L38 and L39 listed, SB-802 and 802.5 HQ fulfill USP L25 requirements, SB-803 HQ fulfills USP L37 requirements, dimensions: 8.0 mm ID x 300 mm L, MW range: 1,000 − 100,000 Daltons in Pullulan
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.