Learn More
Static Solutions Ohm Stat FG-7050 Extra Large Heelgrounder, 1 Meg, 2 Layer
Supplier: Static Solutions FG7050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Ohm-Stat FG-7050 Extra Large Heelgrounder is a 1 megohm 2-layer heel grounder designed to safely drain static electricity from the body. It features a conductive inner layer and an insulating outer layer with an extra large conductive patch for maximum contact area. This durable heel grounder has a 1 megohm resistor to safely drain static charges while preventing shocks. It uses a banana plug for easy attachment to a grounding cord. The FG-7050 provides reliable personal grounding protection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.