Learn More
Static Solutions Ohm Stat FG-7010 D-Ring, 1 Meg, 2 Layer Cup
Supplier: Static Solutions FG7010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Ohm Stat FG-7010 D-Ring 1 Meg 2 Layer Cup is an anti-static cup made of dissipative polyethylene with a surface resistivity of 1 megohm per square. It has a D-ring handle and a 2 layer construction. The cup has a 10 oz capacity and is 3.5" diameter x 3.75" high. It is designed for handling sensitive electrical components that could be damaged by static discharge. The 2 layer construction provides protection from static on both the inside and outside of the cup. The D-ring handle allows for easy handling. With its 1 megohm resistivity, it prevents static buildup while still dissipating charges slowly to protect components.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.