Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Includes 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. Compatible with bellco over head drive an
Supplier: Bellco Glass, Inc. 196436460
Manufactured from type 1 class a Borosilicate glass. Includes 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. Compatible with bellco over head drive an