Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Starter Water Analysis Portable pH Meters

LCD meters combine portability with precise pH measurement.
$393.00 - $525.00
Specifications
Range (pH) | 0 to 14 |
---|---|
Resolution (pH) | 0.01 |
Accuracy (pH) | ±0.01 |
Range (mV) | -1999 to 1999 |
Resolution (mV) | 1mV |
Description
- Auto buffer recognition stores calibration data to avoid errors
- Automatic/manual temperature compensation for accurate readings
- ORP or temperature electrode connectivity for additional functionality
- IP54 protection shields water/dust damage
- Size: 90 × 150 × 35mm
- Weight: 0.18kg
Specifications
0 to 14 | |
±0.01 | |
1mV | |
1 to 3 | |
0.1°C | |
LCD | |
BNC, NTC | |
0.18 kg | |
0.01pH; 1mV; 0.1°C | |
>500 hr. | |
9 cm | |
3.5 cm |
0.01 | |
-1999 to 1999 | |
±1 | |
0°C to 100°C | |
±0.5°C | |
30 Sets, most recent calibration data | |
IP-54 | |
ABS | |
AAA | |
3.5 in. | |
1.3 in. | |
17 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title