Learn More
OHAUS™ Navigator™ NV Portable Balances
With dual multi-functional touchless infrared sensors allowing hands-free operation.
Supplier: OHAUS™ 83033083/EMD
Description
- With EC type approved models
- Ultra-fast response time
- Overload protection for wide variety of weighing applications
- ABS plastic construction with stainless-steel pan
- LCD with backlight display
- Come with front level indicator (lit during power up), non-slip adjustable feet, stability indicator, external menu lock switch, shipping transportation lock, integrated security loop, and superior overload protection
- Six full segment LCD white auto backlight with multiple icons and indicators
- One second stabilization time
Applications
Weighing (14 units), Percentage weighing, Checkweighing, Parts counting, Accumulation/Totalization, and Display hold
Order Info
Units feature mains or battery operation with auto shut-off. Easy to install USB, RS232, and Ethernet interface options.
Certifications
UL listed
Specifications
510 g | |
ABS Plastic, Stainless Steel Pan | |
1 sec. |
0.1 g | |
NV511 | |
Portable Balance |