Learn More
Invivogen ODN 2006 HUM TYPE B CPGS STIM
Supplier: Invivogen TLRL2006
TLR9 Ligands, Stimulatory CpG-ODN Type B, Human Specific, CpG ODNs are Synthetic Oligonucleotides that Contain Unmethylated CpG Dinucleotides in Particular Sequence Contexts (CpG motifs)