Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Ocean Pacific™ Ninja™ Fully Textured Nitrile Medical Examination Gloves

Catalog Number 296367495
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Ocean Pacific™ Ninja™ Fully Textured Nitrile Medical Examination Gloves. This glove provides grip, puncture resistance and protection against bloodborne pathogens, chemical lubricants and sharp instruments. Chemo tested and fentanyl safe. Accepted by Health Canada for food handling.

  • Thick and durable with ample flex for the fine artistry of tattoo, hair design, dentistry, ambulance, laboratory science or pet care.
  • Fully textured to ensure a secure grip, even when wet.
  • Superior puncture resistance provides enhanced protection against bloodborne pathogens, chemical lubricants and sharp instruments.
  • ASTM D6978-05 certified for chemotherapy handling.
  • Evaluated and considered acceptable by Health Canada for food handling.
Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Powdered Powder Free
Size X-Large
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit