Learn More
OccuNomix™ MiraCool™ Pullover Cooling Vest
Click to view available options
Poncho
Description
- 100 percent cotton with super-absorbent cooling crystals
- Works without ice or freezing
- Soak in cold water
- Stays hydrated for days
- Expandable sides with adjustable buckles
- One size fits all
Specifications
Material | Cotton |
Product Type | Pullover Cooling Vest |
For Use With (Application) | For personal cooling |
Quantity | Each |