Learn More
Cole-Parmer Oakton Replacement Turbidity Calibration Solution Kit; 0.02, 20, 100, and 800 NTU Standards
Supplier: Cole-Parmer 3563552
For calibration of Oakton T-100 turbidity meters. Includes one each of 60-mL HPDE bottle of four NTU standards.