Learn More
Cole-Parmer Oakton Replacement Temperature Probe
Supplier: Cole-Parmer 3561313
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Oakton Replacement Temperature Probe, Use With Oakton 700 or 2700 meters, Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample. For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.