Learn More
Cole-Parmer Oakton pH Indicator Strips, 0 to 14, 100/pk
Supplier: Cole-Parmer 3585002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Convenient flip-top vials containing 100 strips include color comparator chart. Read results immediately after exposing test area to the solution. Strips measure 2-3/4"L x 1/4"W (7 x 2 cm).