Learn More
Cole-Parmer Oakton pH 6+ Handheld Meter with pH Probe
Supplier: Cole-Parmer 3561322
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC)ensures accuracy from 0 to 100°C. Meters include a protective rubber boot that also acts as a meter stand.