Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oakton™ Conductivity/TDS/ATC Probes for CON 10, CON 100, CON 200 Meters
Choose from stainless-steel, epoxy and glass bodies; 0.1, 1.0 and 10cm-1 cell constants
Description
For Oakton Conductivity Meters:
- CON 10 (15-500-026)
- CON 100 (15-500-028, 15-500-029)
- CON200 (15-500-030, 15-500-031)
- K= 1.0 for midrange conductivity measurements.
- K= 10.0 for high conductivity measurements near or above 20mS.
- K= 0.1 for low conductivity measurements near or below 20μS
Model Selection:
- Each cell has 3-ft cable and a two-band platinum sensor
- All have built-in temperature compensation
- Epoxy Body cells have a temperature range of 0° to 80°C (32° to 176°F
- Glass Body and Stainless-steel Body cells have a temperature range of 0° to 100°C (32° to 212°F
Specifications
0°C to 80°C | |
0.1 | |
ATC, Conductivity, TDS | |
Epoxy |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction