Learn More
Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Electrical Option
Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.
$743.00 - $1414.00
Specifications
Adjustable | No |
---|---|
Certifications | SEFA compliant and MAS Certified Green™ |
Finish | Chemical resistant, earth-friendly UV |
For Use With (Application) | High school and above and with low temperature use |
Includes | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
S41908
![]() |
Diversified Spaces™
P7304K30E |
24 x 72 in. (60.96 x 182.88 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,019.00
|
|
S41915
![]() |
Diversified Spaces™
P7304K34E |
24 x 72 in. (60.96 x 182.88 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,091.00
|
|
S42107
![]() |
Diversified Spaces™
P7604K30E |
24 x 60 in. (60.96 x 152.4 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $880.00
|
|
S42114
![]() |
Diversified Spaces™
P7604K34E |
24 x 60 in. (60.96 x 152.4 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $952.00
|
|
S42711
![]() |
Diversified Spaces™
P7124K36E |
30 x 48 in. (76.2 x 121.92 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $948.00
|
|
S42121
![]() |
Diversified Spaces™
P7604K36E |
24 x 60 in. (60.96 x 152.4 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $952.00
|
|
S42742
![]() |
Diversified Spaces™
P7144K30E |
30 x 60 in. (76.2 x 152.4 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,075.00
|
|
S47456
![]() |
Diversified Spaces™
P7104K30E |
24 x 48 in. (60.96 x 121.92 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $768.00
|
|
S47466
![]() |
Diversified Spaces™
P7104K36E |
24 x 48 in. (60.96 x 121.92 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $842.00
|
|
S47461
![]() |
Diversified Spaces™
P7104K34E |
24 x 48 in. (60.96 x 121.92 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $842.00
|
|
S47612
![]() |
Diversified Spaces™
P7124K34E |
30 x 48 in. (76.2 x 121.92 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $948.00
|
|
S47607
![]() |
Diversified Spaces™
P7124K30E |
30 x 48 in. (76.2 x 121.92 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $876.00
|
|
S47680
![]() |
Diversified Spaces™
P7134K30E |
30 x 54 in. (76.2 x 137.16 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $990.00
|
|
S47960
![]() |
Diversified Spaces™
P7154K30E |
30 x 72 in. | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,175.00
|
|
S47694
![]() |
Diversified Spaces™
P7134K36E |
30 x 54 in. (76.2 x 137.16 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,064.00
|
|
S47814
![]() |
Diversified Spaces™
P7144K34E |
30 x 60 in. (76.2 x 152.4 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,147.00
|
|
S47821
![]() |
Diversified Spaces™
P7144K36E |
30 x 60 in. (76.2 x 152.4 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,147.00
|
|
S47687
![]() |
Diversified Spaces™
P7134K34E |
30 x 54 in. (76.2 x 137.16 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,064.00
|
|
S47967
![]() |
Diversified Spaces™
P7154K34E |
30 x 72 in. | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,246.00
|
|
S48076
![]() |
Diversified Spaces™
P7164K36E |
21 x 28 in. (53.34 x 121.92 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $814.00
|
|
S48147
![]() |
Diversified Spaces™
P7174K30E |
36 x 48 in. (91.44 x 121.92 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,014.00
|
|
S48069
![]() |
Diversified Spaces™
P7164K34E |
21 x 28 in. (53.34 x 121.92 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $814.00
|
|
S48154
![]() |
Diversified Spaces™
P7174K34E |
36 x 48 in. (91.44 x 121.92 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,084.00
|
|
S48161
![]() |
Diversified Spaces™
P7174K36E |
36 x 48 in. (91.44 x 121.92 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,084.00
|
|
S48233
![]() |
Diversified Spaces™
P7184K34E |
21 x 54 in. (53.34 x 137.16 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $869.00
|
|
S48318
![]() |
Diversified Spaces™
P7194K34E |
36 x 54 in. (91.44 x 137.16 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,188.00
|
|
S48226
![]() |
Diversified Spaces™
P7184K30E |
21 x 54 in. (53.34 x 137.16 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $799.00
|
|
S48311
![]() |
Diversified Spaces™
P7194K30E |
36 x 54 in. (91.44 x 137.16 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,117.00
|
|
S48240
![]() |
Diversified Spaces™
P7184K36E |
21 x 54 in. (53.34 x 137.16 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $869.00
|
|
S48325
![]() |
Diversified Spaces™
P7194K36E |
36 x 54 in. (91.44 x 137.16 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,188.00
|
|
S42784
![]() |
Diversified Spaces™
P7164K30E |
21 x 28 in. (53.34 x 121.92 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $743.00
|
|
S48436
![]() |
Diversified Spaces™
P7204K34E |
24 x 54 in. (60.96 x 137.16 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $924.00
|
|
S48429
![]() |
Diversified Spaces™
P7204K30E |
24 x 54 in. (60.96 x 137.16 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $853.00
|
|
S48556
![]() |
Diversified Spaces™
P7214K30E |
21 x 60 in. (53.34 x 152.4 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $854.00
|
|
S48443
![]() |
Diversified Spaces™
P7204K36E |
24 x 54 in. (60.96 x 137.16 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $924.00
|
|
S48563
![]() |
Diversified Spaces™
P7214K34E |
21 x 60 in. (53.34 x 152.4 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $924.00
|
|
S48570
![]() |
Diversified Spaces™
P7214K36E |
21 x 60 in. (53.34 x 152.4 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $924.00
|
|
S41812
![]() |
Diversified Spaces™
P7244K30E |
36 x 72 in. (91.44 x 182.88 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,341.00
|
|
S48641
![]() |
Diversified Spaces™
P7224K30E |
36 x 60 in. (91.44 x 152.4 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,214.00
|
|
S48655
![]() |
Diversified Spaces™
P7224K36E |
36 x 60 in. (91.44 x 152.4 cm) | 36 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,285.00
|
|
S48648
![]() |
Diversified Spaces™
P7224K34E |
36 x 60 in. (91.44 x 152.4 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,285.00
|
|
S48709
![]() |
Diversified Spaces™
P7234K30E |
21 x 72 in. (53.34 x 182.88 cm) | 30 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $972.00
|
|
S41819
![]() |
Diversified Spaces™
P7244K34E |
36 x 72 in. (91.44 x 182.88 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,414.00
|
|
S48716
![]() |
Diversified Spaces™
P7234K34E |
21 x 72 in. (53.34 x 182.88 cm) | 34 in. | Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only |
Each for $1,043.00
|
|
Description
- 1 in. Phenolic Table Surface
- 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
- Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
- Diversified Spaces™ is SEFA compliant and MAS Certified Green
- Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
- Includes UL rated electrical that can be hard wired or portable cord connected by certified electrician
- Limited Lifetime Warranty
- Made in the USA
- Minimal assembly required; legs need to be attached
- Solid Oak Construction
Specifications
No | |
Chemical resistant, earth-friendly UV | |
Electrical Cutout, Box, Duplex Receptacle, and Cover Plate Only | |
Plain Apron | |
500 lb. | |
226.79 kg |
SEFA compliant and MAS Certified Green™ | |
High school and above and with low temperature use | |
Red Oak | |
Solid Phenolic | |
Plain Apron Table |