Learn More
Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top
Ideal for high school and above with lower temperature use.
$708.00 - $1614.00
Specifications
Certifications | MAS Certified Green™ |
---|---|
Finish | Chemical resistant, earth-friendly UV |
For Use With (Application) | High school and above with lower temperature use |
Frame Material | Red Oak |
Height (English) | 30 in. |
Alert:
Do not use with high temperatures. Exposed
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
S41911
![]() |
Diversified Spaces™
P7304K30N |
24 x 72 in. (60.96 x 182.88 cm) |
Each for $984.00
|
|
S42020
![]() |
Diversified Spaces™
P7484K30N |
48 in. dia. |
Each for $1,614.00
|
|
S42222
![]() |
Diversified Spaces™
P7804K30L |
42 x 54 in. (106.68 x 137.16 cm) |
Each for $1,194.00
|
|
S42280
![]() |
Diversified Spaces™
P7904K30L |
42 x 60 in. (106.68 x 152.4 cm.) |
Each for $1,307.00
|
|
S42591
![]() |
Diversified Spaces™
P7404K30L |
42 x 72 in. (106.68 x 182.88 cm) |
Each for $1,456.00
|
|
S42603
![]() |
Diversified Spaces™
P7604K30N |
24 x 60 in. (60.96 x 152.4 cm) |
Each for $844.00
|
|
S42706
![]() |
Diversified Spaces™
P7124K30N |
30 x 48 in. (76.2 x 121.92 cm) |
Each for $841.00
|
|
S47459
![]() |
Diversified Spaces™
P7104K30N |
24 x 48 in. (60.96 x 121.92 cm) |
Each for $734.00
|
|
S47546
![]() |
Diversified Spaces™
P7114K30L |
42 x 48 in. (106.68 x 121.92 cm) |
Each for $1,094.00
|
|
S47683
![]() |
Diversified Spaces™
P7134K30N |
30 x 54 in. (76.2 x 137.16 cm) |
Each for $956.00
|
|
S47810
![]() |
Diversified Spaces™
P7144K30N |
30 x 60 in. (76.2 x 152.4 cm) |
Each for $1,040.00
|
|
S47963
![]() |
Diversified Spaces™
P7154K30N |
30 x 72 in. (76.2 x 182.88 cm.) |
Each for $1,138.00
|
|
S48065
![]() |
Diversified Spaces™
P7164K30N |
21 x 48 in. (53.34 x 121.92 cm) |
Each for $708.00
|
|
S48150
![]() |
Diversified Spaces™
P7174K30N |
36 x 48 in. (91.44 x 121.92 cm) |
Each for $978.00
|
|
S48229
![]() |
Diversified Spaces™
P7184K30N |
21 x 54 in. (53.34 x 137.16 cm) |
Each for $765.00
|
|
S48314
![]() |
Diversified Spaces™
P7194K30N |
36 x 54 in. (91.44 x 137.16 cm) |
Each for $1,082.00
|
|
S48432
![]() |
Diversified Spaces™
P7204K30N |
24 x 54 in. (60.96 x 137.16 cm) |
Each for $817.00
|
|
S41815
![]() |
Diversified Spaces™
P7244K30N |
36 x 72 in. (91.44 x 182.88 cm) |
Each for $1,306.00
|
|
S48644
![]() |
Diversified Spaces™
P7224K30N |
36 x 60 in. (91.44 x 152.4 cm) |
Each for $1,180.00
|
|
S48712
![]() |
Diversified Spaces™
P7234K30N |
21 x 72 in. (53.34 x 182.88 cm) |
Each for $938.00
|
|
S48559
![]() |
Diversified Spaces™
P7214K30N |
21 x 60 in. (53.34 x 152.4 cm) |
Each for $820.00
|
|
Description
- Solid red oak legs and apron
- Black rubber leg boots
- Chemical-resistant, earth-friendly UV finish
Specifications
MAS Certified Green™ | |
High school and above with lower temperature use | |
30 in. | |
Non-skid adjustable glides and rubber leg boots | |
Solid Phenolic | |
226.79 kg |
Chemical resistant, earth-friendly UV | |
Red Oak | |
76.2 cm | |
Red Oak | |
500 lb. |