Learn More
Beckman Coulter Replacement O-Ring for Canister Closure 70mm ID, 12/pk
Supplier: Beckman Coulter 360848
Replacement O-ring for the canister closure of the JLA-10.500 Rotor (package of 12) and compatible with canisters with closures (part # 360828).