Learn More
Chemglass Life Sciences O-Ring, Viton, #341
Supplier: Chemglass Life Sciences CG305341
O-Ring, Viton, #341. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200º C. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.