Learn More
iQ Biosciences New Zealand White Rabbit Splenocytes, 25M cells/vial
Supplier:  iQ Biosciences IQBRBSP103
Encompass_Preferred
High-quality, high-viability (typically > 70%) splenocytes from the New Zealand white (NZW) rabbit, a commonly-used outbred rabbit strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2855620