Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Nylon Test Tube Brush
With fan-shaped end.
Supplier: Eisco™ CH0204GPK12
Description
- Made from nylon
- Wire-wound handle with loop
Specifications
Tube Brush | |
Stainless Steel Wire | |
White | |
230 mm |
Nylon | |
Fan Tip | |
20 mm | |
6 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction