Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Nylon Cord
High quality, coated Nylon Cord offers reduced friction and is typically used during pulley and pendulum demonstrations.
Supplier: Eisco™ PH0302A
Description
- Coated to reduce friction
- Cord is shipped rolled on a cardboard cone
- Approximate Length: 30 m
Specifications
Coated Nylon | |
Cord | |
30 m |
Pulley and Pendulum Demonstrations | |
98.4 ft. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction