Learn More
Cosmo Bio Usa Inc Nycodenz AG, 500G
Supplier: Cosmo Bio Usa Inc AXS1002424
Nycodenz is a universal density gradient medium provided as an non-ionic, non-toxic powder that is freely soluble in water. Solutions up to 80% (v/w) with a density of 1.426 g/ml can be prepared. It is a tri-iodinated derivative of benzoic acid with three aliphatic hydrophilic side chains. Nycodenz is the ideal medium for:Mammalian and non-mammalian cellsSubcellular organellesOrganelles from nonmammalian sourcesSubcellular membranesProtein and protein complexes
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.