Learn More
LGC Biosearch Technologies NxGen T7 RNA PolymeraseReliable and robust enzyme for production of mRNA transcripts in a variety of applications.
Supplier: LGC Biosearch Technologies 302231
NxGen T7 RNA PolymeraseReliable and robust enzyme for production of mRNA transcripts in a variety of applications.Supplied at 50,000 U/mL in 50 mM Tris-HCl, 100 mM NaCl, 1 mM dithiothreitol, 1 mM EDTA, 50% glycerol, 0.1% Triton X-100, pH 7.9 @ 25°C. Also provided is 10X T7 RNA Polymerase Buffer which in 1X form is composed of 40 mM Tris-HCl, 6 mM MgCl2, 10 mM Dithiothreitol, 2 mM Spermidine, pH 7.9 @ 25°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.