Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Nutrient Agar
High quality microbiological media at an economical price
Supplier: Innovating Science™ MD0100CS/24
This item is not returnable.
View return policy
Description
Specifications
Sterile | |
Room Temperature | |
United States | |
Prepared Microbiology Media | |
Microbes, Educational | |
200 mL |
1 | |
Bottle | |
Nutrient Agar | |
Microbes | |
Glass |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction