Learn More
Cytiva NSB reducer, 10mL
Supplier: Cytiva BR100691
Contents: Carboxymethyl dextran sodium salt (10 mg/ml) in 0.15 M MaCl containing 0.02% sodium azide (NaN3), 10 ml; Additive to reduce non-specific binding in dextran-based sensor surfaces when running complex sample matrices in Biacore systems. Reduces nonspecific binding to carboxymethyl dextran sensor surfaces. Sufficient for approximately 650 samples.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.