Learn More
Yamato Scientific America, Inc. Yamato 150L Forced Convection Oven, Energy saving, 220V
Supplier: Yamato Scientific America, Inc. DNE611
Energy saving, programmable 150L forced convection eco-oven with lower power consumption and CO2 emission. High precision controller allows high performance temperature control and display of CO₂ and power discharge. Program operation with a maximum of 99 steps, 99 patterns repeatable. Standard equipped with various support functions such as calibration offset, power failure recovery mode, save and access of user setting information, as well as other operation modes. Data acquisition from internal test device possible because of cable holes. Easy system upgrade with various option settings. Operating temp ambient +20~210°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.